Skip to Content
Merck
All Photos(1)

Key Documents

ABC116

Sigma-Aldrich

Anti-EPS8, Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Epidermal growth factor receptor kinase substrate 8, EPS8

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€344.00

€344.00


Estimated to ship on10 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€344.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€344.00


Estimated to ship on10 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

species reactivity (predicted by homology)

human (immunogen homology), rat (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... EPS8(2059)

General description

EPS8 (epidermal growth factor receptor kinase substrate 8), which was originally identified as a substrate for the kinase activity of EGFR, is a multifunctional protein that is known to play a role in spermatogenesis, the regulation of actin, and other cellular processes. EPS8 uses its SH3 domain to complex with Abi1 and SOS1, leading to Rac activation and subsequent actin cytoskeletal remodeling. In the same manner, it binds with RN-tre to inhibit the internalization of EGFR. EPS8 also works with Arp3 to modulate actin dynamics within the seminiferous epithelium at the apical ES and the BTB.

Immunogen

KLH-conjugated linear peptide corresponding to human EPS8.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This EPS8 antibody is validated for use in WB for the detection of the EPS8 protein.

Quality

Evaluated by Western Blot in NIH/3T3 cell lysate.

Western Blot Analysis: 0.2 µg/mL of this antibody detected EPS8 in 10 µg of NIH/3T3 cell lysate.

Target description

~97 kDa observed. A second isoform is known to exist at ~68 kDa Uncharacterized bands may be observed at ~50 kDa and ~37 kDa in some cell lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
NIH/3T3 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service