Skip to Content
Merck
All Photos(1)

Key Documents

AB9712

Sigma-Aldrich

Anti-Sortilin Antibody

serum, Chemicon®

Synonym(s):

Neurotensin receptor 3

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
€401.00

€401.00


Estimated to ship on21 March 2025Details


Request a Bulk Order

Select a Size

Change View
50 μL
€401.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€401.00


Estimated to ship on21 March 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SORT1(6272)

General description

Neurotensin receptor 3 or sortilin (the intracellular sorting protein 95 kDa) is a member of a family of receptors characterized by a single transmembrane domain, a short intracytoplasmic domain, and a long cysteine-rich luminal extracellular domain. This receptor has been found in the brain, spinal cord, muscle and other peripheral cell types, and in cancer cells. Sortilin exhibits sequence similarities with proteins involved in intracellular transport and trafficking.

Specificity

Sortilin (Neurotensin receptor 3). By Western blot the antibody recognizes a strong band at ~90 kDa in extracts of NSC34 cells. By immunohistochemistry the antibody intensely stains motor neurons within the rat spinal cord.

Immunogen

Recombinant protein from the extracellular domain of human sortilin.

Application

Anti-Sortilin Antibody is an antibody against Sortilin for use in WB, IH.
Western blot: 1:1,000-1:2,000 on extracts from NSC34 cells.
Immunohistochemistry: 1:500-1:1,000 on rat spinal cord.

Optimal working dilutions must be determined by the end user.

Physical form

Lyophilized. Reconstitute with 50 μL of sterile distilled water. Centrifuge to remove any insoluble material.

Storage and Stability

Maintain lyophilized material at -20 to -70°C for up to 12 months after date of receipt. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. Glycerol (ACS grade or better) can be added 1:1 for additional stability.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kimberly S Williams et al.
Journal of neuroimmunology, 285, 76-93 (2015-07-23)
To characterize the role of neurotrophin receptors on macrophages, we investigated the ability of nerve growth factor (NGF) and its precursor, proNGF, to regulate human macrophage phenotype. The p75 neurotrophin receptor (p75(NTR)) and TrkA were concentrated within overlapping domains on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service