Skip to Content
Merck
All Photos(1)

Key Documents

AB9676

Sigma-Aldrich

Anti-Tau phospho Threonine 205 Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€587.00

€587.00


Estimated to ship on28 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€587.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€587.00


Estimated to ship on28 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pThr205)

Gene Information

human ... MAPT(4137)

General description

Tau is a neuronal microtubule-associated protein found predominantly on axons and functions to promote tubulin polymerization and stabilize microtubules. Tau, in its hyperphosphorylated form, is the major component of paired helical filaments (PHF), the building block of neurofibrillary lesions in Alzheimer′s disease (AD) brain. Hyperphosphorylated Tau is also found in neurofibrillary lesions in a range of other central nervous system disorders. Hyperphosphorylation impairs the microtubule binding function of Tau, resulting in the destabilization of microtubules in AD brains, ultimately leading to the degeneration of the affected neurons. Numerous serine/threonine kinases, including GSK-3beta, protein kinase A (PKA), cyclin-dependent kinase 5 (cdk5) and casein kinase II (CK2), phosphorylate Tau. Phosphorylation of threonine 205 is catalyzed GSK-3beta and cdk5 in vitro.

Specificity

Tau phosphoThreonine 205. The antibody recognizes Tau pThreonine 205 in samples of recombinant human Tau treated with GSK-3beta for 45 minutes.. The reactivity of the antibody is blocked with the pThreonine 205 peptide but not the non-phosphopeptide or a generic phosphoThreonine-containing peptide.

Immunogen

Synthetic peptide of amino acids surrounding the phosphoThreonine 205 site of human Tau.

Application

Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases
This Anti-Tau phospho Threonine 205 Antibody is validated for use in WB for the detection of Tau phospho Threonine 205.
Western blot: 1:1,000. Suggested blocking buffer is 5% BSA-TBST for one hour at room temperature. Suggested antibody dilution buffer is 3% BSA-TBST. Suggested antibody incubation time is 2 hours at room temperature..


Optimal working dilutions must be determined by the end user.

Storage and Stability

Maintain at -20°C in undiluted for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles. Do not store in a self defrosting freezer.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service