Skip to Content
Merck
All Photos(1)

Key Documents

AB9630

Sigma-Aldrich

Anti-NMDAR2B phospho Tyr1252 Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€287.00

€287.00


Estimated to ship on05 May 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€287.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€287.00


Estimated to ship on05 May 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pTyr1252)

Gene Information

Specificity

NMDAR2B, phosphoTyrosine1252. The antibody recognizes a protein of ~180 kDa corresponding to NMDAR2B, phosphoTyrosine1252 in lysates from rat brain hippocampus. Immunolabeling is blocked by the phosphopeptide used as the immunogen but not the corresponding non-phosphopeptide. Immunolabeling is also blocked by Lamda-phosphatase treatment.
The immunogen has 100% homology with human and non-human primates.

Immunogen

Synthetic peptide of amino acids surrounding the phosphoTyrosine 1252 site of rat NMDAR2B.

Application

Anti-NMDAR2B phospho Tyr1252 Antibody is an antibody against NMDAR2B phospho Tyr1252 for use in WB.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service