Skip to Content
Merck
All Photos(1)

Key Documents

AB9328

Sigma-Aldrich

Anti-Thioredoxin 1 Antibody

serum, Chemicon®

Synonym(s):

TRX1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€335.00

€335.00


Estimated to ship on25 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€335.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€335.00


Estimated to ship on25 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat

species reactivity (predicted by homology)

human (sequence homology 14/15)

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... TXN(7295)

Specificity

Recognizes Thioredoxin 1 (TRX1).

Immunogen

Synthetic peptide from rat/mouse Thioredoxin 1.

Application

Detect Thioredoxin 1 using this Anti-Thioredoxin 1 Antibody validated for use in WB.
Immunoblotting: 1:1,000-1:5,000 using ECL. Reacts with the ~12-14 kD thioredoxin 1 protein. Additional bands >30 kD may been seen.

Suggested dilution and blocking buffer is TBST containing 5% non-fat milk. Suggested gel percentage is 15%. Suggested incubation time is overnight.
Research Category
Neuroscience
Research Sub Category
Oxidative Stress

Linkage

Replaces: MABN1090

Physical form

Rabbit serum. Liquid. Contains no preservative.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoqin Zhao et al.
Acta biochimica et biophysica Sinica, 46(4), 318-329 (2014-03-01)
Cardiac complications are the leading cause of death in diabetes. However, the mechanism of diabetes in inducing myocardial injury and apoptosis, and whether the thioredoxin (Trx) system is involved remain unclear. In this study, male Sprague-Dawley rats were randomly divided
Chiara Milanese et al.
Nature communications, 10(1), 4887-4887 (2019-10-28)
Accumulation of DNA lesions causing transcription stress is associated with natural and accelerated aging and culminates with profound metabolic alterations. Our understanding of the mechanisms governing metabolic redesign upon genomic instability, however, is highly rudimentary. Using Ercc1-defective mice and Xpg

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service