Skip to Content
Merck
All Photos(1)

Key Documents

AB5927

Sigma-Aldrich

Anti-Acid Sensitive Ion Channel 3 Antibody, 285-304 of extracellular domain of rat ASIC3

Chemicon®, from guinea pig

Synonym(s):

ASIC3, Amiloride-sensitive Cation Channel 3, TNaC1, Testis Sodium Channel 1, Dorsal Root Ganglion ASIC (DRASIC)

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
€622.00

€622.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
50 μL
€622.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€622.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

biological source

guinea pig

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Specificity

ASIC3 (DRASIC)

Immunogen

Epitope: 285-304 of extracellular domain of rat ASIC3
Synthetic peptide from the extracellular domain of rat ASIC3 (Catalog Number AG366). (C)TASLDPDDFDPEPSDPLGSP

Application

Detect Acid Sensitive Ion Channel 3 using this Anti-Acid Sensitive Ion Channel 3 Antibody, 285-304 of extracellular domain of rat ASIC3 validated for use in IC, IH.
Immunohistochemistry: 1:100

Immunocytochemistry: 1:100

Optimal working dilutions must be determined by the end user.

APPLICATION NOTES FOR AB5927

IMMUNOHISTOCHEMISTRY

Male Sprague-Dawley rats (b.wt. 100-150g) were anesthetized with sodium pentobarbital and perfused via the ascending aorta with: 1) 50 mL of Ca2+-free Tyrode+s solution followed by 2) a formalin-picric acid fixative (4% paraformaldehyde with 0.4% picric acid in 0.16 M phosphate buffer, pH 6.9) and 3) 10% sucrose in PBS as a cryo-protectant. Tissues were rapidly dissected out and stored overnight in 0.1 M phosphate buffer (pH 7.4) containing 10% sucrose. Slide-mounted tissue sections were incubated with blocking buffer for 1 hour at room temperature. Primary antibody was diluted in blocking buffer to the appropriate working dilution. Blocking buffer was removed and the slides were then incubated at 4°C for 18-24 hours with AB5927. After rinsing in PBS 3 times sections were incubated for 60 minutes at room temperature with Cy3-conjugated secondary antibodies. After mounting in a mixture of PBS and glycerol (1:3) containing 0.1% p-phenylenediamine, sections were examined with a Nikon Microphot-SA epifluorescence microscope.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters

Neuroinflammation & Pain

Physical form

Affinity purified immunoglobulin. Liquid in PBS, pH 7.4 with 1% BSA and 0.05% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted for up to 6 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mathieu Meleine et al.
Scientific reports, 10(1), 9986-9986 (2020-06-21)
Ghrelin is the only orexigenic peptide currently known and a potent prokinetic by promoting gastric motility but novel insights suggest that its role extends beyond satiety regulation. Whereas ghrelin was shown to provide somatic and colonic antinociception, its impact on
Age-dependent sensitization of cutaneous nociceptors during developmental inflammation.
Jankowski, MP; Ross, JL; Weber, JD; Lee, FB; Shank, AT; Hudgins, RC
Molecular Pain null
Tanguy Chaumette et al.
Pain, 161(5), 1109-1123 (2020-01-25)
Mechanical allodynia is a cardinal sign of several inflammatory pain disorders where nerve growth factor, a prototypic neurotrophin, plays a crucial role by binding to TrkA receptors. Here, we took the advantage of our generated knock-in mouse model expressing a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service