Skip to Content
Merck
All Photos(2)

Key Documents

AB5873-I

Sigma-Aldrich

Anti-Catechol O-methyltransferase/COMT Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Catechol O-methyltransferase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€317.00

€317.00


Estimated to ship on17 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€317.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€317.00


Estimated to ship on17 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

bovine (based on 100% sequence homology), rhesus macaque (based on 100% sequence homology), horse (based on 100% sequence homology), porcine (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... COMT(1312)

General description

Catechol O-methyltransferase (EC 2.1.1.6; UniProt P21964) is encoded by the COMT gene (Gene ID 1312) in human. Catechol-O-methyltransferase initiates the metabolic breakdown of catecholamines (including the neurotransmitters dopamine, epinephrine, and norepinephrine) by catalyzing the transfer of a methyl group from S-adenosylmethionine (SAM) to catecholamines in the presence Mg++. This O-methylation results in one of the major degradative pathways of the catecholamine transmitters. In addition to endogenous substances, COMT is also important in the metabolism of catechol drugs. Two isoforms exist as a result of alternative initiation codon usage. The membrane-bound isoform or MB-COMT contains a short N-terminal cytoplasmic tail (a.a. 1-6), a transmembrane segment (a.a. 7-26), and a large C-terminal extracellular region (a.a. 27-271), while the soluble or S-COMT isoform lacks the N-terminal 50-amino acid sequence of MB-COMT. MB-COMT and S-COMT differ in their substrate affinity and capacities. Despite having a lower capacity than S-COMT, MB-COMT has around a 10-fold greater affinity for dopamine and noradrenaline, and therefore is likely to be better suited for metabolizing catecholamines at the concentrations found in brain. The influence of the Val(158)Met polymorphic form of catechol-o-methyltransferase (COMT) on brain activation and functional connectivity is the subject of numerous studies.

Specificity

This polyclonal antibody was raised against a sequence present in both the membrane-bound (MB-COMT) and the soluble (S-COMT) spliced isoforms of human catechol O-methyltransferase reported by UniProt (P21964).

Immunogen

Epitope: Extracellular domain.
KLH-conjugated linear peptide corresponding to an extracellular domain sequence of human catechol O-methyltransferase.

Application

Immunohistochemistry Analysis: A 1:2,000 dilution from a representative lot detected Catechol O-methyltransferase/COMT in human prostate cancer, normal liver and kidney tissue sections.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
This Anti-Catechol O-methyltransferase Antibody is validated for use in Western Blotting, Immunohistochemistry for the detection of COMT.

Quality

Evaluated by Western Blotting in human fetal liver tissue lysate.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected Catechol O-methyltransferase/COMT in 10 µg of human fetal liver tissue lysate.

Target description

~28 kDa observed. 30.04 kDa (human MB-COMT) and 24.45 kDa (human S-COMT) calculated. Uncharacterized band(s) may appear in some lysates.

Linkage

Replaces: AB5873

Physical form

Affinity purified.
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Taiga Miyajima et al.
Free radical biology & medicine, 147, 69-79 (2019-12-21)
Fuchs Endothelial Corneal Dystrophy (FECD) is an age-related genetically complex disease characterized by increased oxidative DNA damage and progressive degeneration of corneal endothelial cells (HCEnCs). FECD has a greater incidence and advanced phenotype in women, suggesting a possible role of
Varun Kumar et al.
Redox biology, 69, 102986-102986 (2023-12-14)
Fuchs endothelial corneal dystrophy (FECD) is a genetically complex, age-related, female-predominant disorder characterized by loss of post-mitotic corneal endothelial cells (CEnCs). Ultraviolet-A (UVA) light has been shown to recapitulate the morphological and molecular changes seen in FECD to a greater

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service