Skip to Content
Merck
All Photos(1)

Key Documents

AB5850

Sigma-Aldrich

Anti-GABA B Receptor R1 Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€410.00

€410.00


Estimated to ship on28 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€410.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€410.00


Estimated to ship on28 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

spider, rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

Specificity

GABAB Receptor 1 subunit, N-terminus.

Immunogen

amino acid 33-54 of the rat GABA(B)R1 protein (PHLPRPHPRVPPHPSSERRAVY)

Application

Immunoblotting: 1:500-1:1,000. The antibody should detect a ~106kDa band under standard western conditions.

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
This Anti-GABA B Receptor R1 Antibody is validated for use in WB for the detection of GABA B Receptor R1.

Physical form

Format: Purified
Purified immunoglobulin. Liquid. Buffer = PBS containing 0.02% sodium azide.

Storage and Stability

Maintain frozen at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qi Zhang et al.
Neural plasticity, 2018, 8919347-8919347 (2018-10-27)
Electroacupuncture (EA) has been reported to benefit hypertension, but the underlying mechanisms are still unclear. We hypothesized that EA attenuates hypertension, in part, through modulation of γ-aminobutyric acid (GABA) receptor function in the nucleus tractus solitarii (NTS). In the present

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service