Skip to Content
Merck
All Photos(1)

Key Documents

AB5675-AF488

Sigma-Aldrich

Anti-Metabotropic Glutamate Receptor 5 Antibody, Alexa Fluor 488

from rabbit, ALEXA FLUOR 488

Synonym(s):

Metabotropic glutamate receptor 5, mGluR5, G protein-coupled receptor GRM5, G protein coupled receptor family C, group 1, member E

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€512.00

€512.00


Estimated to ship on08 May 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€512.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€512.00


Estimated to ship on08 May 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

species reactivity (predicted by homology)

bovine (based on 100% sequence homology), primate (based on 100% sequence homology), horse (based on 100% sequence homology), human (based on 100% sequence homology), mouse (based on 100% sequence homology), feline (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GRM5(2915)

General description

Metabotropic glutamate receptor 5 or mGluR5 (UniProt P31424; also known as G protein-coupled receptor GRM5, G protein coupled receptor family C, group 1, member E) is encoded by the Grm5 (also known as Gprc1e, Mglur5) gene (Gene ID 24418) in rat species. The metabotropic glutamate receptors belong to a family of G protein-coupled receptors that have been divided into 3 groups based on their sequence homology, putative signal transduction mechanisms, and pharmacologic properties. mGluR1 and mGluR5 constitute Group I metabotropic glutamate receptors. mGluR5 is found on postsynaptic terminals of neurons and microglia. mGluR5 signals via Gq/G11 coupling to activate phospholipase C, resulting in calcium mobilization and activation of protein kinase C (PKC). In addition, mGluR5 is also reported to mediate signaling involving Src-family protein tyrosine kinase in a G protein-independent manner.

Specificity

Expected to react with all spliced isoforms of human, rat, and mouse mGluR5 reported by UniProt (P41594, P31424 and Q3UVX5, respectively).

Immunogen

Epitope: C-terminal cyoplasmic domain.
Synthetic peptide corresponding to a sequence from the C-terminal cytoplasmic domain of rat mGluR5.

Application

Anti-Metabotropic Glutamate Receptor 5 Antibody, Alexa Fluor 488 is an antibody against Metabotropic Glutamate Receptor 5 for use in Immunocytochemistry, Immunofluorescence.
Immunocytochemistry Analysis: An 1:100 dilution from a representative lot detected Metabotropic Glutamate Receptor 5 in rat cortical neurons.
The unconjugated antibody (Cat. No. AB5675) is shown to be suitable also for Western blotting and immunohistochemistry applications.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Quality

Evaluated by Immunofluorescence in rat brain tissue sections.

Immunofluorescence Analysis: An 1:100 dilution of this antibody detected Metabotropic Glutamate Receptor 5 in rat cerebellum and rat brain tissue sections.

Target description

~150 kDa observed.

Physical form

Affinity purified
Purified rabbit polyclonal antibody conjugate in PBS with 15 mg/mL BSA and 0.1 % sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service