Skip to Content
Merck
All Photos(1)

Key Documents

AB5589

Sigma-Aldrich

Anti-Potassium Channel Kv1.3 Antibody, extracellular domain

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
€1,150.00

€1,150.00


Estimated to ship on15 April 2025Details


Request a Bulk Order

Select a Size

Change View
200 μL
€1,150.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€1,150.00


Estimated to ship on15 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KCNA3(3738)

Specificity

Recognizes Kv1.3, extracellular (Kcna3).

Immunogen

Epitope: extracellular domain
Highly purified peptide corresponding to residues 263-275 of human Kv1.3 (Accession P22001).

Application

Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
This Anti-Potassium Channel Kv1.3 Antibody, extracellular domain is validated for use in FC, WB for the detection of Potassium Channel Kv1.3.
Western blotting: 1:200-1:500 using ECL on rat brain membranes.

Indirect flow cytometry: 0.5-5 μg of antibody per 1x10e6cells.

Dilutions should be made using a carrier protein such as BSA (1-3%).

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Lyophilized from PBS, pH 7.4, containing 1% BSA and 0.05% sodium azide. Reconstitute with 200 μL of sterile distilled water. Centrifuge antibody preparation before use (10,000 x g for 5 min).

Storage and Stability

Maintain lyophilized material at -20°C for up to 6 months after date of receipt. After reconstitution maintain antibody and control peptide at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
Included free of charge with the antibody is 40 μg of control antigen (lyophilized powder). For negative control, preincubate 5 μg of purified peptide with 1 μg of antibody for one hour at room temperature. Optimal concentrations must be determined by the end user.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luiz O Leiria et al.
The Journal of physiology, 591(9), 2259-2273 (2013-03-13)
We aimed to investigate the role of insulin in the bladder and its relevance for the development of overactive bladder (OAB) in insulin-resistant obese mice. Bladders from male individuals who were involved in multiple organ donations were used. C57BL6/J mice
Ameet A Chimote et al.
The Journal of biological chemistry, 287(3), 2055-2067 (2011-12-03)
Hypoxia in solid tumors contributes to decreased immunosurveillance via down-regulation of Kv1.3 channels in T lymphocytes and associated T cell function inhibition. However, the mechanisms responsible for Kv1.3 down-regulation are not understood. We hypothesized that chronic hypoxia reduces Kv1.3 surface
Zerrin Kuras et al.
PloS one, 7(8), e43859-e43859 (2012-09-07)
The migration of T lymphocytes is an essential part of the adaptive immune response as T cells circulate around the body to carry out immune surveillance. During the migration process T cells polarize, forming a leading edge at the cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service