Skip to Content
Merck
All Photos(2)

Key Documents

AB5421-I

Sigma-Aldrich

Anti-Vesicular Glutamate Transporter 3 (VGLUT3) Antibody

serum, from guinea pig

Synonym(s):

Vesicular glutamate transporter 3, Solute carrier family 17 member 8, VGluT3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€388.00

€388.00


Estimated to ship on02 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€388.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€388.00


Estimated to ship on02 April 2025Details


Request a Bulk Order

biological source

guinea pig

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human, rat

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

General description

Vesicular glutamate transporter 3 (UniProt Q7TSF2; also known as Solute carrier family 17 member 8, VGluT3) is encoded by the Slc17a8 (also known as Vglut3) gene (Gene ID 266767) in rat species. Vesicular glutamate transporters (VGluTs) are proton-dependent carriers responsible for the upload of the major excitatory neurotransmitter glutamate into synaptic vesicles within presynaptic terminals to allow later regulated release at the synaptic cleft. Studies conducted in mice show that VGluT1 and VGluT2 are essential for vital functions, while Vglut3-knockout mice have a normal life expectancy despite being deaf and developing anxiety and discrete basal locomotor phenotypes. VGluT1- and VGluT2-expressing neurons represent the bulk of canonical glutamatergic neurons in the brain, with VgluT1 representing the major subtype and accounting for approximately 80% of total vesicular transport of glutamate in the brain. In contrast, VgluT3 is found in discrete population of non-glutamatergic neurons using other transmitters than glutamate, such as cholinergic interneurons in the dorsal and ventral striatum, subpopulations of GABAergic basket cells in the cortex and hippocampus, and serotoninergic neurons, where it is involved in more subtle modulation of local transmission. Rat VGluT3 is a 12-transmembrane (a.a. 77-97, 131-151, 154-174, 183-203, 222-242, 250-270, 315-335, 354-374, 391-411, 414-434, 448-468, 482-502) protein with 7 cytoplasmic and 6 vacuolar domains, having both its N- and C-terminal tails (a.a. 1-76 and 503-585) exposed at the cytoplasmic side.

Specificity

This polyclonal guinea pig antiserum was raised against a VGluT3 C-terminal cytoplasmic domain sequence conserved between rat and murine species and shares 96% homology with the corresponding human sequence.

Immunogen

Epitope: C-terminal cytoplasmic domain.
KLH-conjugated linear peptide corresponding to a C-terminal cytoplasmic domain sequence of rat vesicular glutamate transporter 3 (VGluT3).

Application

Anti-Vesicular Glutamate Transporter 3 (VGLUT3) is an antibody against Vesicular Glutamate Transporter 3 (VGLUT3) for use in Immunocytochemistry and Western Blotting.
Immunocytochemistry Analysis: A 1:250 dilution from a representative lot detected vesicular glutamate transporter 3 (VGluT3) in PC12 rat pheochromocytoma cells.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Quality

Evaluated by Western Blotting in PC12 cell lysate.

Western Blotting Analysis: A 1:2,000 dilution of this antibody detected vesicular glutamate transporter 3 (VGluT3) in 10 µg of PC12 rat pheochromocytoma cell lysate.

Target description

~61 kDa observed. 64.76 kDa (rat), 66.15 kDa (mouse), and 64.99/59.08 kDa (human isoform 1/2) calculated. A ~130 kDa central nervous system VGluT3 isoform has also been described (Munguba, G.C., et al. (2011). Mol. Vis. 17:413-419). Uncharacterized band(s) may appear in some lysates.

Linkage

Replaces: AB5421

Physical form

Guinea pig polyclonal antibody serum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiadong Xu et al.
Scientific reports, 11(1), 15779-15779 (2021-08-06)
Haplo-insufficiency of the GATA3 gene causes hypoparathyroidism, sensorineural hearing loss, and renal disease (HDR) syndrome. Previous studies have shown that Gata3 is required for the development of the prosensory domain and spiral ganglion neurons (SGNs) of the mouse cochlea during
Nandkishore Prakash et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 40(3), 569-584 (2019-12-04)
Chronic stress induces anhedonia in susceptible but not resilient individuals, a phenomenon observed in humans as well as animal models, but the molecular mechanisms underlying susceptibility and resilience are not well understood. We hypothesized that the serotonergic system, which is
Ke Ning et al.
Investigative ophthalmology & visual science, 62(9), 15-15 (2021-07-10)
Primary cilia are conserved organelles found in polarized cells within the eye that regulate cell growth, migration, and differentiation. Although the role of cilia in photoreceptors is well-studied, the formation of cilia in other retinal cell types has received little

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service