Skip to Content
Merck
All Photos(1)

Key Documents

AB5154

Sigma-Aldrich

Anti-Cav2.2 Antibody

Chemicon®, from rabbit

Synonym(s):

Voltage Gated Calcium Channel (N-type) alpha 1B Subunit, Cacna1b

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
€777.00

€777.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

Select a Size

Change View
200 μL
€777.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€777.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (tissue)
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

Specificity

Recognizes a 1B subunit (both LMW and HMW forms) of voltage-gated calcium channel. Does not cross react with any other calcium channel antigens tested so far.

SPECIES REACTIVITIES: It is expected that the antibody will also work on mouse due to sequence homology (15/17). Other species have not been tested.

Immunogen

Purified peptide CNB1 from 851-867 of alpha 1B subunit of rat brain voltage-gated calcium channel (Accession Q02294; Westenbroek et al. 1992) containing N-terminal lysine and tyrosine, conjugated to KLH with gluteraldehyde (Harlow & Lane 1988).

Application

Detect Cav2.2 using this Anti-Cav2.2 Antibody validated for use in IC, IP & WB.
In preparing full-length protein samples for Western blot or immunoprecipitation, the procedures should be performed at 4°C, with the following protease inhibitor mixture: pepstatin A (1 μg/mL), leupeptin (1 μg/mL), aprotinin (1 μg/mL), Pefabloc SC (0.2 mM), benzamidine (0.1 mg/mL), and calpain inhibitors I and II (8 μg/mL each).

Immunoprecipitation

Immunocytochemistry. Recommend subsequent staining with a labeled second antibody.

Western blot: 1.5 to 3.0 μg/mL (1:100-1:200) using ECL

Dilutions should be made using a carrier protein such as BSA (1-3%)

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters

Neuroinflammation & Pain

Physical form

Affinity purified immunoglobulin. Lyophilized from PBS, pH 7.4, containing 1% BSA and 0.05% sodium azide as a preservative. Reconstitute with 200 μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 xg for 5 min).

Storage and Stability

Maintain lyophilized material at -20°C for up to 12 months after date of receipt. After reconstitution maintain at 2-8°C in undiluted aliquots for up to 2 weeks. For longer periods, it is recommended to add glycerol (ACS grade or better) at a ratio of 1:1 and store the antibody at -20°C for up to 6 months. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
Included free of charge with the antibody is 40 μg of control antigen (lyophilized powder). The stock solution of the antigen can be made up using 100 μL of sterile deionized water. For negative control, preincubate 1 μg of peptide with 10 μg of antibody for one hour at room temperature. Optimal concentrations must be determined by the end user.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J H Xu et al.
Neuropathology and applied neurobiology, 36(1), 71-85 (2009-10-09)
To investigate the subcellular localization of Ca(v)2.2 calcium channel in the mouse central nervous system (CNS), and changes of Ca(v)2.2 at acute and chronic stages during and after pilocarpine-induced status epilepticus (PISE), in order to find out the roles it
Lisa M Smyth et al.
Neuropharmacology, 56(2), 368-378 (2008-10-01)
Using HPLC techniques we evaluated the electrical field stimulation-evoked overflow of noradrenaline (NA), adenosine 5'-triphosphate (ATP), and beta-nicotinamide adenine dinucleotide (beta-NAD) in the presence of low nanomolar concentrations of omega-conotoxin GVIA or omega-agatoxin IVA in the canine mesenteric arteries and
Kang Zhang et al.
Frontiers in pharmacology, 8, 302-302 (2017-06-13)
The sigma-1 receptor is a 223 amino acids molecular chaperone with a single transmembrane domain. It is resident to eukaryotic mitochondrial-associated endoplasmic reticulum and plasma membranes. By chaperone-mediated interactions with ion channels, G-protein coupled receptors and cell-signaling molecules, the sigma-1
Geraldine A Fuller-Bicer et al.
American journal of physiology. Heart and circulatory physiology, 297(1), H117-H124 (2009-05-12)
Cardiac L-type voltage-dependent Ca(2+) channels are heteromultimeric polypeptide complexes of alpha(1)-, alpha(2)/delta-, and beta-subunits. The alpha(2)/delta-1-subunit possesses a stereoselective, high-affinity binding site for gabapentin, widely used to treat epilepsy and postherpetic neuralgic pain as well as sleep disorders. Mutations in
Luca Trovò et al.
EMBO reports, 25(6), 2610-2634 (2024-05-03)
GABAB receptors (GBRs), the G protein-coupled receptors for GABA, regulate synaptic transmission throughout the brain. A main synaptic function of GBRs is the gating of Cav2.2-type Ca2+ channels. However, the cellular compartment where stable GBR/Cav2.2 signaling complexes form remains unknown.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service