Skip to Content
Merck
All Photos(1)

Key Documents

AB3549P

Sigma-Aldrich

Anti-Monocarboxylate Transporter 8 Antibody

Chemicon®, from rabbit

Synonym(s):

MCT8, X-linked PEST-containing Transporter

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€335.00

€335.00


Estimated to ship on02 May 2025Details


Request a Bulk Order

Select a Size

Change View
50 μG
€335.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€335.00


Estimated to ship on02 May 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SLC16A2(6567)

General description

MCT8 is an integral membrane protein that is associated with the plasma membrane. A member of the Major Facilitator Superfamily, MCT8 serves as a proton-linked monocarboxylate transporter that catalyzes the rapid transport of monocarboxylates and ketone bodies across the plasma membrane. Expression of MCT8 is greatest in liver and kidney.

Specificity

Recognizes mouse Monocarboxylate Transporter 8.

Immunogen

A 19 amino acid peptide sequence within the cytoplasmic, C-terminus of mouse MCT8. The immunogen sequence is 94% conserved in rat and 89% in human MCT8 and shows no significant sequence homology with other MCT family members.

Application

Anti-Monocarboxylate Transporter 8 Antibody is an antibody against Monocarboxylate Transporter 8 for use in ELISA & WB.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
Western blot: 1-5 μg/mL using ECL.
ELISA: 1:10,000-1:100,000 using 50-100 ng control peptide (Catalog number AG202) per well.

Optimal working dilutions must be determined by the end user.

Linkage

Replaces: AB3324P

Physical form

Affinitypurified immunoglobulin presented as a liquid in PBS with 0.1% BSA.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months from date of receipt. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
POSITIVE CONTROL: Liver or kidney extracts

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service