Skip to Content
Merck
All Photos(1)

Key Documents

AB3365P

Sigma-Aldrich

Anti-Resistin-like Molecule α Antibody

Chemicon®, from rabbit

Synonym(s):

RELMalpha

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€336.00

€336.00


Estimated to ship on21 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€336.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€336.00


Estimated to ship on21 March 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Retnla(57262)

Specificity

Reacts with mouse RELM-alpha expressed by E. coli or by 293 cells (RELM-alpha-Fc). Recombinant mouse RELM-alpha forms mono and dimmer on protein gel.

Immunogen

Full length recombinant mouse RELM-alpha.

Application

Anti-Resistin-like Molecule α Antibody is an antibody against Resistin-like Molecule α for use in ELISA & WB.
Research Category
Inflammation & Immunology
Research Sub Category
Cytokines & Cytokine Receptors
Western Blot

ELISA

Optimal working dilutions must be determined by the end user.

Physical form

Format: Purified
Purified immunoglobulin. Liquid.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

During shipment, small volumes of product will occasionally become entrapped in the seal of the product vial. For products with volumes of 200 μL or less, we recommend gently tapping the vial on a hard surface or briefly centrifuging the vial in a tabletop centrifuge to dislodge any liquid in the container′s cap.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edson Kiyotaka Ishizuka et al.
International immunopharmacology, 14(4), 513-522 (2012-09-04)
Recently our group described that Nattectin, a C-type lectin of the venom of Thalassophryne nattereri shows a potent pro-inflammatory capacity. Here, we demonstrated that Nattectin is able to induce M1 macrophage marker iNOS, and up-regulate the expression of MHC class

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service