Skip to Content
Merck
All Photos(1)

Key Documents

AB3006

Sigma-Aldrich

Anti-CREB Antibody

Chemicon®, from rabbit

Synonym(s):

CREB-1, cAMP-response Element-binding Protein-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€358.00

€358.00


Estimated to ship on11 February 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€358.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€358.00


Estimated to ship on11 February 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
electrophoretic mobility shift assay: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CREB1(1385)

General description

CREB is a beta ZIP transcription factor that activates target genes through cAMP response elements. CREB is able to mediate signals from numerous physiological stimuli, resulting in regulation of a broad array of cellular responses. While CREB is expressed in numerous tissues, it plays a large regulatory role in the nervous system. CREB is believed to play a key role in promoting neuronal survival, precursor proliferation, neurite outgrowth and neuronal differentiation in certain neuronal populations. Additionally, CREB signaling is involved in learning and memory in several organisms. CREB is able to selectively activate numerous downstream genes through interactions with different dimerization partners. CREB is activated by phosphorylation at Ser133 by various signaling pathways including Erk, Ca2+ and stress signaling. Some of the kinases involved in phosphorylating CREB at Ser133 are p90RSK, MSK, CaMKIV and MAPKAPK-2.

Specificity

Human cyclic AMP responsive element binding protein-1 (CREB-1, p43). By Western blot it shows a major band at 46 kDa when assayed against a lysate of 1 x 106 Raji B cells. Minor bands at 75 kDa and 43 kDa may be noted. Partial reactivity to CREM-1 protein. May react non-specifically with other proteins.

Immunogen

CREB-1 (p43) peptide corresponding to the N-terminal (aa 5-45) of human protein conjugated to KLH.

Application

Immunoblot

Immunoprecipitation

ELISA

Supershift assay: 0.5-1.0 μL per assay.

Optimal dilutions must be determined by the end user.
Use Anti-CREB Antibody (Rabbit Polyclonal Antibody) validated in ELISA, EMSA, IP, WB to detect CREB also known as CREB-1, cAMP-response Element-binding Protein-1.

Linkage

Replaces: 04-218

Physical form

Format: Purified
Purified immunoglobulin. Liquid (sterile filtered) in 0.02M Potassium Phosphate buffer containing 0.15 M Sodium Chloride, pH 7.2 with 0.01% sodium azide.

Storage and Stability

Maintain frozen at -20°C in undiluted aliquots for up to six months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maike Stegen et al.
PloS one, 16(2), e0247087-e0247087 (2021-02-19)
G protein-coupled receptor kinase 6 (GRK6) is part of the G protein-coupled receptor kinase family, whose members act as key regulators of seven-transmembrane receptor signalling. GRK6 seems to play a role in regulation of inflammatory processes, but mechanisms of transcriptional
Ling-Ling Teng et al.
PLoS biology, 17(3), e2007097-e2007097 (2019-03-19)
Dietary restriction (DR; sometimes called calorie restriction) has profound beneficial effects on physiological, psychological, and behavioral outcomes in animals and in humans. We have explored the molecular mechanism of DR-induced memory enhancement and demonstrate that dietary tryptophan-a precursor amino acid
Jonathan Aryeh Sobel et al.
PLoS biology, 15(4), e2001069-e2001069 (2017-04-18)
Many organisms exhibit temporal rhythms in gene expression that propel diurnal cycles in physiology. In the liver of mammals, these rhythms are controlled by transcription-translation feedback loops of the core circadian clock and by feeding-fasting cycles. To better understand the
Diego Scheggia et al.
Brain, behavior, and immunity, 97, 89-101 (2021-07-12)
Autoantibodies targeting the GluA3 subunit of AMPA receptors (AMPARs) have been found in patients with Rasmussen's encephalitis and different types of epilepsy and were associated with the presence of learning and attention deficits. Our group recently identified the presence of
Dynamic histone H3 acetylation and methylation at human cytomegalovirus promoters during replication in fibroblasts.
Cuevas-Bennett, C; Shenk, T
Journal of virology null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service