Skip to Content
Merck
All Photos(1)

Key Documents

AB22-2ML

Sigma-Aldrich

Goat Anti-Human IgG Antibody, heavy and light chains

serum, Chemicon®

Synonym(s):

Goat Anti-Human IgG, Goat IgG Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

2 ML
€139.00

€139.00


Estimated to ship on22 April 2025Details



Select a Size

Change View
2 ML
€139.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

€139.00


Estimated to ship on22 April 2025Details


biological source

goat

Quality Level

antibody form

serum

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunoprecipitation (IP): suitable

shipped in

dry ice

target post-translational modification

unmodified

Related Categories

Specificity

Goat Anti-Human IgG secondary antibody recognizes Human IgG (H & L). May show reactivity to immunoglobulins from other species.

Immunogen

Epitope: heavy and light chains
Human IgG, whole molecule.

Application

Goat anti-Human IgG Antibody, heavy & light chains is an antibody against Human IgG for use in IP.
Precipitation: 1:15-1:40.

Final working dilutions must be determined by end user.
Research Category
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies

Physical form

Goat serum. Liquid, PBS 0.01M, 0.05% Sodium Azide, pH 7.1

Storage and Stability

Maintain lyophilized material at 2-8ºC for up to one year. After reconstitution maintain at -20°C in undiluted aliquots for up to six months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Review the key factors that should figure in your decision to choose a secondary antibody. Learn about species, subclass, isotype, label, and more.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service