Skip to Content
Merck
All Photos(1)

Key Documents

AB1926

Sigma-Aldrich

Anti-Integrin beta 5 (ITGB5) Antibody

CHEMICON®, rabbit polyclonal

Synonym(s):

Anti-Anti-AA475909, Anti-Anti-AI874634, Anti-Anti-ESTM23, Anti-Anti-[b]-5, Anti-Anti-[b]5, Anti-Anti-[b]5A, Anti-Anti-[b]5B, Anti-Anti-beta-5, Anti-Anti-beta5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€421.00

€421.00


Estimated to ship on28 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€421.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€421.00


Estimated to ship on28 April 2025Details


Request a Bulk Order

Product Name

Anti-Integrin β5 Antibody, CT/cytosolic, serum, Chemicon®

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, hamster, human, rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
radioimmunoassay: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... ITGB5(3693)
mouse ... Itgb5(16419)
rat ... Itgb5(257645)

General description

The integrin family of cell adhesion receptors consists of at least 16 membrane-associated heterodimers, composed of an alpha and beta subunit that associate in a non-covalent manner. The structure and functional diversity of the integrin family are based upon the pairing abilities of the individual alpha and beta subunits. Key to these molecular interactions between the integrin receptors and their respective ligands is the recognition of the Arg-Gly-Asp (RGD) sequence, known to be present in the extracellular matrix components fibronectin, vitronectin, collagen, fibrinogen, and von Willebrand factor (Cheresh, 1991). The involvement of integrins in vascular proliferation, adhesion, and wound repair has been well documented.

The beta5 subunit associates with alphaV forming a receptor for vitronectin. This complex is expressed in a wide range of cell types including fibroblast and keratinocytes. It is also expressed at high levels in several tumors including lung, pancreatic and mammary carcinomas. It is not expressed in lymphblastoid cells.

Specificity

Recognizes Integrin beta5. Specificity determined by immunoprecipitation from detergent extract of 35S methionine labeled cells. No cross-reactivity to beta1 or beta3.

Immunogen

Epitope: C-terminus/cytosolic
Synthetic peptide derived from the COOH terminal sequence (cytoplasmic domain) of the human integrin beta 5 subunit (KTFNKFNKSYNGTVD). Note : a lysine (K) residue was added to the N-terminal of the peptide to facilitate coupling to the carrier protein.

Application

Detect Integrin β5 using this Anti-Integrin β5 Antibody, C-terminus/cytosolic validated for use in ELISA, IF, IH(P), IP, RIA & WB.
ELISA/RIA

Immunoprecipitation: 5μL of antibody is sufficient to precipitate alphaVbeta5 from

2.5x10E6 cells.

Western blot: non-reduced conditions; do not use milk in the dilution buffer.

Immunofluorescence: 1:100.

Immunohistochemistry (Acetone Fixed) 1:40-1:80

Immunohistochemistry (Paraffin Embedded) 1:200 (ABC)

Optimal working dilutions must be determined by end user.

Target description

88 kDa

Physical form

UnPurified whole rabbit Serum containing 0.05% sodium azide as a preservative.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Syndecan-1 regulates alphavbeta5 integrin activity in B82L fibroblasts.
McQuade, KJ; Beauvais, DM; Burbach, BJ; Rapraeger, AC
Journal of Cell Science null
Mechanical strain regulates osteoblast proliferation through integrin-mediated ERK activation.
Yan, YX; Gong, YW; Guo, Y; Lv, Q; Guo, C; Zhuang, Y; Zhang, Y; Li, R; Zhang, XZ
Testing null
Integrins control dendritic spine plasticity in hippocampal neurons through NMDA receptor and Ca2+/calmodulin-dependent protein kinase II-mediated actin reorganization.
Shi, Y; Ethell, IM
The Journal of Neuroscience null
J Watzlawik et al.
Glia, 58(15), 1782-1793 (2010-07-21)
Human remyelination promoting IgM mAbs target oligodendrocytes (OLs) and function in animal models of multiple sclerosis (MS). However, their mechanism of action is unknown. This study seeks to identify the cellular mechanism of action of a recombinant human IgM on
Molecular consequences of silencing mutant K-ras in pancreatic cancer cells: justification for K-ras-directed therapy.
Fleming, JB; Shen, GL; Holloway, SE; Davis, M; Brekken, RA
Molecular Cancer Research null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service