Skip to Content
Merck
All Photos(1)

Key Documents

AB1922

Sigma-Aldrich

Anti-Integrin β4 Antibody

serum, Chemicon®

Synonym(s):

CD104

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€390.00

€390.00


Estimated to ship on30 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€390.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€390.00


Estimated to ship on30 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGB4(3691)
mouse ... Itgb4(192897)
rat ... Itgb4(25724)

Specificity

Specific for beta4 integrin as determined by immunoprecipitation. The epitope recognized is on the cytoplasmic domain of the protein and is thus intracellular. No cross-reactivity to other integrin b subunits.

Immunogen

Synthetic peptide derived from the COOH terminal sequence (cytoplasmic domain) of the human β4 integrin subunit (Tamura et al. 1990). The sequence used is NH2-(K)GTLSTHMDQQFFQT-amide. The lysine (K) is not in the β4 sequence and was added to conjugate the peptid

Application

Anti-Integrin β4 Antibody is an antibody against Integrin β4 for use in IP, IH.
Immunoprecipitation from labeled cell extracts: 5 μL of serum is sufficient to precipitate alpha6beta4 from 1-2 x 10E6 cells. In some cases immunoprecipitation from 35S methionine labeled cells results in two bands of 95 and 55 kDa of unknown origin in addition to the 200 kDa and 140 kDa bands corresponding to beta4 and alpha6.

Immunohistochemistry: 5 μg/mL. See protocol below.

Optimal working dilutions must be determined by the end user.



Suggested Protocol for Immunohistochemical:

1. Fix cryostat sections with a 1:1 chloroform-acetone mixture for 10 minutes at room temperature.

2. Air Dry.

3. Wash with 2 mL of 1x PBS.

4. Add 200 μL of first antibody (5 μg/mL) to each slide. Incubate for 1 hour at room temperature in a humidity chamber.

5. Wash with 5 mL of 1x PBS.

6. Add appropriately diluted anti-rabbit secondary antibody conjugate and incubate for 1 hour in a humidity chamber

7. Wash with 5 mL of 1x PBS.

8. Add 200 μL of development solution (containing DAB or other substrate depending on the secondary antibody) and incubate at room temperature for the necessary time to develop the dark color. Follow the reaction with a microscope, to establish the correct color intensity.

9. Block the reaction by washing with 2 mL of 1x PBS.

10. Counterstain with hematoxylin as desired.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palmitoylation by DHHC3 is critical for the function, expression, and stability of integrin ?6?4.
Sharma, C; Rabinovitz, I; Hemler, ME
Cellular and Molecular Life Sciences null
Haiyao Wang et al.
The Journal of biological chemistry, 290(43), 26103-26113 (2015-09-10)
The α6β4 integrin is known to associate with receptor tyrosine kinases when engaged in epithelial wound healing and in carcinoma invasion and survival. Prior work has shown that HER2 associates with α6β4 integrin and syndecan-1 (Sdc1), in which Sdc1 engages
Haiyao Wang et al.
The Journal of biological chemistry, 289(44), 30318-30332 (2014-09-10)
Epithelial cells are highly dependent during wound healing and tumorigenesis on the α6β4 integrin and its association with receptor tyrosine kinases. Previous work showed that phosphorylation of the β4 subunit upon matrix engagement depends on the matrix receptor syndecan (Sdc)-1
K Triantafilou et al.
Journal of virology, 74(13), 5856-5862 (2000-06-14)
Human parechovirus 1 (HPEV1) displays an arginine-glycine-aspartic acid (RGD) motif in the VP1 capsid protein, suggesting integrins as candidate receptors for HPEV1. A panel of monoclonal antibodies (MAbs) specific for integrins alphavbeta3, alphavbeta1, and alphavbeta5, which have the ability to
Expression of laminin isoforms, receptors, and binding proteins unique to nucleus pulposus cells of immature intervertebral disc.
Chen, J; Jing, L; Gilchrist, CL; Richardson, WJ; Fitch, RD; Setton, LA
Connective tissue research null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service