Skip to Content
Merck
All Photos(1)

Key Documents

AB10627

Sigma-Aldrich

Anti-Omentin Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€347.00

€347.00


Estimated to ship on02 May 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€347.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€347.00


Estimated to ship on02 May 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITLN1(55600)

General description

A calcium-dependent lectin with affinity for furanosides, such as galactofuranosyl in bacterial arabinogalactans. 30kD of apparent molecular weight of Omentin migrate at 40-kD under reducing conditions due to glycosylation. Omentin, a novel visceral fat depot-specific secretory protein, enhance insulin-stimulated glucose transport in subcutaneous as well as omental adipocytes and may therefore be implicated in the pathogenesis of obesity.

Specificity

Recognizes Omentin in western blotting.

Immunogen

Epitope: Amino acids 95-115 of Human Omentin.
Synthetic peptide: TVGDRWSSQQGSKADYPEGDG, corresponding to amino acids 95-115 of Human Omentin.

Application

Anti-Omentin Antibody detects level of Omentin & has been published & validated for use in ELISA & WB.
Western blot: 1:2,000-1:5,000 using ECL. Reacted with the ~40kDa Omentin His-tagged protein. Suggested blocking and dilution buffer is PBST with 0.05% Tween-20 and 5% skim milk. Suggested incubation time is 1 hour at room temperature.

ELISA, direct and indirect: 1:2,000-1:5,000.

Target description

30-40 kDa

Physical form

Format: Purified
Liquid in PBS, pH7.4. Contains no preservatives.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kseniia V Dracheva et al.
Non-coding RNA, 9(6) (2023-11-21)
Obesity is a significant risk factor for the development of type 2 diabetes mellitus (T2DM). Adipose tissue dysfunction can affect the pool of circulating exosomal miRNAs, driving concomitant disease in obesity. These exosomal miRNAs can reflect adipose tissue functionality, thus
Chi-Lam Au-Yeung et al.
Nature communications, 11(1), 3546-3546 (2020-07-17)
Advanced ovarian cancer usually spreads to the omentum. However, the omental cell-derived molecular determinants modulating its progression have not been thoroughly characterized. Here, we show that circulating ITLN1 has prognostic significance in patients with advanced ovarian cancer. Further studies demonstrate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service