Skip to Content
Merck
All Photos(1)

Key Documents

AB10626

Sigma-Aldrich

Anti-IL-1 β Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
€377.00

€377.00


Estimated to ship on07 May 2025Details


Request a Bulk Order

Select a Size

Change View
500 μG
€377.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€377.00


Estimated to ship on07 May 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

swine, pig

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

General description

This antibody recognizes natural and recombinant swine IL-1 . either a 269 AA or a 271 AA (17 kDa) protein with no disulfide bonds. There are two distinct molecular forms of IL-1 (IL-1 and IL-1 ) derived from two different genes.

Specificity

Other species were not tested.
Recognizes IL-1 Beta.

Immunogen

Recombinant swine IL-1β.

Application

Anti-IL-1 β Antibody is an antibody against IL-1 β for use in WB.
Research Category
Signaling
Research Sub Category
Growth Factors & Receptors
This antibody is suitable for use in Western blot analysis at 0.5 - 2.0 ug/mL.
This antibody is non-neutralizing when tested by proliferation assay.

Linkage

Replaces: 04-462; MABF22

Physical form

Format: Purified
Protein G Purified
Rabbit polyclonal antibodies in phosphate buffered saline, pH 7.2.

Storage and Stability

Store at -20°C in undiluted aliquots for 1 year from date of shipment.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuan Lu et al.
Frontiers in pharmacology, 11, 703-703 (2020-06-17)
Dysfunction of human endothelial cells is an important trigger for atherosclerosis. Oxidative low-density lipoprotein (ox-LDL) usually was used to stimulate the dysfunction of human umbilical vein endothelial cells (HUVECs). LncRNA SNHG1 (small nucleolar RNA host gene 1) is a cerebral
Influences of lamin A levels on induction of pluripotent stem cells.
Zuo, B; Yang, J; Wang, F; Wang, L; Yin, Y; Dan, J; Liu, N; Liu, L
Biology Open null
Xiaojuan Han et al.
Redox biology, 44, 102010-102010 (2021-06-04)
Activated microglia are an important type of innate immune cell in the brain, and they secrete inflammatory cytokines into the extracellular milieu, exert neurotoxicity to surrounding neurons and are involved in the pathogenesis of many brain disorders. Quercetin (Qu), a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service