Skip to Content
Merck
All Photos(1)

Key Documents

AB10305

Sigma-Aldrich

Anti-FXR-B Antibody

serum, Chemicon®

Synonym(s):

Bile acid receptor, Farnesoid X-activated receptor, Farnesol receptor HRR-1, Nuclear receptor subfamily 1 group H member 4, Retinoid X receptor-interacting protein 14, RXR-interacting protein 14

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NR1H4(9971)

General description

The farnesoid X receptor (FXR) is a member of the nuclear receptor superfamily and is known to play a key role in bile acid homeostasis. It does through the regulation of key genes involved in bile acid synthesis, metabolism and transport.

Immunogen

Recombinant human FXR-B

Application

Suggested dilutions:
Western Blotting: 1:5,000
Detect FXR-B using this Anti-FXR-B Antibody validated for use in WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

Evaluated by Western Blotting of recombinant human FXR-B.

Western Blotting Analysis: A 1:5,000 dilution of this rabbit polyclonal antiserum detected recombinant human FXR-B.

Target description

~56 kDa observed.

Physical form

Rabbit polyclonal antiserum with 0.1% sodium azide.

Storage and Stability

Maintain at -20°C for up to 12 months from date of receipt.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service