Skip to Content
Merck
All Photos(1)

Key Documents

9Q078W

Sigma-Aldrich

Anti-canine NKp46 (CD335) Antibody, clone 48A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€362.00

€362.00


Estimated to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€362.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€362.00


Estimated to ship on31 March 2025Details


Request a Bulk Order

antibody form

purified antibody

clone

48A
monoclonal

purified by

using protein G

species reactivity

canine, mouse

concentration

(Please refer to lot specific datasheet.)

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable

isotype

IgG2aκ

UniProt accession no.

General description

L-cells expressing canine NKp46 fusion protein

Specificity

Clone 48A is a mouse monoclonal antibody that specifically detects canine NKp46.

Application

Epitope: extracellular domain

Quality

Evaluated by Immunocytochemistry in canine peripheral blood mononuclear cells (PBMCs).

Immunocytochemistry Analysis: A 1:250 dilution of this antibody detected canine NKp46 (CD335) in canine peripheral blood mononuclear cells (PBMCs).

Physical properties

36.13 kDa calculated.

Physical form

Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Canine.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service