Skip to Content
Merck
All Photos(1)

Key Documents

946627

Supelco

Hand pump ball

for use with solvent withdrawal systems

Synonym(s):

red pump ball, pump ball with connector

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€443.00

€443.00


Estimated to ship on27 March 2025Details


Request a Bulk Order

Select a Size

Change View
1 PKG
€443.00

About This Item

UNSPSC Code:
41122409
NACRES:
NB.51

€443.00


Estimated to ship on27 March 2025Details


Request a Bulk Order

compatibility

for use with solvent withdrawal systems

shipped in

ambient

storage temp.

no temp limit

Related Categories

General description

If withdrawal system parts are lost or damaged,  spare parts are available, e.g. hand pump balls for our withdrawal systems. This pump ball is equipped with a rapid-action connector for the manual withdrawal system 1.01123.0001.
with rapid-action connector

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service