Skip to Content
Merck
All Photos(1)

Documents

9210-OP

Millipore

Tris Base

≥99.8%, needles, crystalline powder, OmniPur® Grade

Synonym(s):

Tris(hydroxymethyl)aminomethane, Tris Base, Tris(Hydroxymethyl)aminomethane, Tris(hydroxymethyl)aminomethane, 2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris), Trizma, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Trometamol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(CH2OH)3
CAS Number:
Molecular Weight:
121.14
Beilstein:
741883
MDL number:
UNSPSC Code:
12161700
NACRES:
NA.21

product name

TRIS, OmniPur® Grade, >=99.8%

Quality Level

form

crystalline powder
needles

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

useful pH range

7-9

pKa (25 °C)

8.1

bp

219-220 °C/10 mmHg (lit.)

mp

167-172 °C (lit.)

shipped in

ambient

storage temp.

room temp

SMILES string

NC(CO)(CO)CO

InChI

1S/C4H11NO3/c5-4(1-6,2-7)3-8/h6-8H,1-3,5H2

InChI key

LENZDBCJOHFCAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Suitable for a wide range of molecular biology applications. Molecular biology grade.

Specifications
DNAse: none detected
RNAse: none detected
Protease: none detected
Arsenic (As): ≤0.0005%
Copper (Cu): ≤0.0001%
Iron (Fe): ≤0.0001%
Lead (Pb): ≤0.0001%
Magnesium (Mg): ≤0.0005%
Loss on drying: ≤1%
A280 (1 M, water): ≤0.05 AU
Purity: ≥99.8% by assay
Solubility: 10% in water
Insoluble matter: ≤0.005%





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Warning

Toxicity: Irritant (B)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service