Skip to Content
Merck
All Photos(1)

Key Documents

8.57006

Sigma-Aldrich

Capping Reagent B2 for DNA Synthesis

60% Lutidine in MeCN (v/v), Novabiochem®

Synonym(s):

60% Lutidine in MeCN (v/v), Novabiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
€200.00
4 L
€1,710.00

€200.00


Estimated to ship on26 May 2025Details


Request a Bulk Order

Select a Size

Change View
500 ML
€200.00
4 L
€1,710.00

About This Item

UNSPSC Code:
12352401
NACRES:
NA.22

€200.00


Estimated to ship on26 May 2025Details


Request a Bulk Order

Quality Level

product line

Novabiochem®

form

liquid

reaction suitability

reaction type: Acetylations
reagent type: derivatization reagent

manufacturer/tradename

Novabiochem®

transition temp

flash point 10.0 °C

density

0.87 g/cm3 at 20 °C

storage temp.

2-30°C

General description

Pre-mixed ready-to-use Capping Reagent
  • 857006.0500: Compatible with Cytiva ÄKTA DNA Synthesisers. Attaches directly to Instrument (GL-45 Neck Finish).

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

50.0 °F

Flash Point(C)

10.0 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service