Skip to Content
Merck
All Photos(1)

Key Documents

8.55126

Sigma-Aldrich

NovaPEG amino resin

Novabiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51171641

product line

NovaPEG
Novabiochem®

Quality Level

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

amine

storage temp.

2-8°C

General description

A remarkable totally PEG-based resin for solid phase peptide synthesis. The support has excellent swelling properties in a wide range of solvents, including water, MeCN, MeOH, DCM, DMF, THF, and toluene. In comparative studies this resin was found to give better results than polystyrene-based supports in the synthesis of hydrophobic peptides. The high hydrophilicity was also found to benefit on-resin immunoassays with one-bead-one-peptide libraries [1].

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Literature references

[1] F. Garcia-Martin, et al. (2006) J. Comb. Chem., 8, 213.

Linkage

Replaces: 01-64-0485

Analysis Note

Color (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (determined from the substitution of the Fmoc-Leu loaded resin): 0.45 - 1.00 mmol/g
TNBS-Test: passes test
Swelling Volume (in DMF): lot specific result

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service