Skip to Content
Merck
All Photos(1)

Key Documents

8.53118

Sigma-Aldrich

Trt-Cys(Trt)-OH.DEA Novabiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

8531180005
€72.30

€72.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
8531180005
€72.30

About This Item

UNSPSC Code:
12352401
NACRES:
NA.22

€72.30


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service