Skip to Content
Merck
All Photos(1)

Key Documents

8.52375

Sigma-Aldrich

Fmoc-hLys(Boc)-OH

Novabiochem®

Synonym(s):

Fmoc-hLys(Boc)-OH, N-α-Fmoc-N-ω-t.-Boc-L-homolysine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€814.00

€814.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€814.00

About This Item

Empirical Formula (Hill Notation):
C27H34N2O6
CAS Number:
Molecular Weight:
482.57
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

€814.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

product line

Novabiochem®

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

amine

storage temp.

2-8°C

SMILES string

N([C@@H](CCCCCNC(=O)OC(C)(C)C)C(=O)O)C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C27H34N2O6/c1-27(2,3)35-25(32)28-16-10-4-5-15-23(24(30)31)29-26(33)34-17-22-20-13-8-6-11-18(20)19-12-7-9-14-21(19)22/h6-9,11-14,22-23H,4-5,10,15-17H2,1-3H3,(H,28,32)(H,29,33)(H,30,31)/t23-/m0/s1

InChI key

CLFMIWUWBOOJLY-QHCPKHFHSA-N

General description

This homolog of lysine has been used to modulate ion-pairing interactions between Lys and enzyme carboxylate groups in SAR studies [1].

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Literature references

[1] K. J. Kennedy, et al. (1997) Bioorg. Med. Chem. Lett., 7, 1937.

Application

Fmoc-hLys(Boc)-OH can be used as a building block in the solid phase peptide synthesis.[1][2]

Analysis Note

Colour (visual): white to off white
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 99.5 % (a/a)
Purity (TLC(CMA2)): ≥ 97 %
Assay (HPLC, area%): ≥ 95.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Probing the flexibility of the catalytic nucleophile in the lyase catalytic pocket of human DNA polymerase β with unnatural lysine analogues
Daskalova SM, et al.
Biochemistry, 56(3), 500-513 (2017)
Exploring the impact of the side-chain length on peptide/RNA binding events
Sbicca L, et al.
Physical Chemistry Chemical Physics, 19(28), 18452-18460 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service