Skip to Content
Merck
All Photos(1)

Key Documents

8.52340

Sigma-Aldrich

N-Biotinyl-NH(PEG)11-COOH

Novabiochem®

Synonym(s):

N-Biotinyl-NH(PEG)11-COOH, O-[2-(Biotinylamino)ethyl]-OÆ-(2-carboxyethyl)undecaethylene glycol

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€504.00
1 G
€1,530.00

€504.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€504.00
1 G
€1,530.00

About This Item

Empirical Formula (Hill Notation):
C37H69N3O16S
Molecular Weight:
844.02
UNSPSC Code:
12352106
NACRES:
NA.22

€504.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

product line

Novabiochem®

Assay

≥98% (TLC)

form

powder

reaction suitability

reaction type: Biotinylations

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

storage temp.

15-25°C

General description

The use of this novel biotin derivative incorporates a hydrophilic 40 atom PEG spacer between the peptide and biotin. This not only greatly improves the solubility of the resultant peptide, but reduces steric hindrance between the peptide and biotin, leading to better avidin binding and higher biological activity. The biotin-PEG derivative can be easily incorporated using any standard coupling method as the final step in SPPS, immediately prior to cleavage of the peptide from the resin.

Associated Protocols and Technical Articles
Biotinylation Reagents for Peptide Synthesis

Analysis Note

Colour (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Purity (TLC(CMA2)): ≥ 98 %
Identity (LC-MS): 843.0 - 845.0 g/mol

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service