Skip to Content
Merck
All Photos(1)

Key Documents

8.52113

Sigma-Aldrich

Fmoc-Asp(biotinyl-PEG)-OH

Novabiochem®

Synonym(s):

Fmoc-Asp(biotinyl-PEG)-OH, N-α-Fmoc-N-γ-(N-biotinyl-3-(2-(2-(3-aminopropyloxy)-ethoxy)-ethoxy)-propyl)-L-asparagine

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€444.00
1 G
€797.00

€444.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€444.00
1 G
€797.00

About This Item

Empirical Formula (Hill Notation):
C39H53N5O10S
Molecular Weight:
783.93
UNSPSC Code:
12352209
NACRES:
NA.22

€444.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

product line

Novabiochem®

Assay

≥95.0% (HPLC)
≥98% (TLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

biotin

storage temp.

2-8°C

General description

In contrast to Fmoc-Lys(biotin)-OH, this novel biotin-labeled amino acid has excellent solubility in DMF and other solvents used in Fmoc SPPS [1]. The PEG-spacer restricts hindrance between the peptide and avidin, leading to better biotin binding. Furthermore, the hydrophilic nature of the PEG minimizes non-specific interactions that can arise from the spacer group becoming buried in the hydrophobic pocket of proteins.

Associated Protocols and Technical Articles
Biotinylation Reagents for Peptide Synthesis

Literature references

[1] B. Baumeister, et al. (2003) Biopolymers, 71, 339.

Linkage

Replaces: 04-12-1279

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 99.5 % (a/a)
Purity (TLC(CMA2)): ≥ 98 %
Assay (HPLC, area%): ≥ 95.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Biotin-labelled peptides find applications in immunology and histochemistry for affinity purification and receptor localization.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service