Skip to Content
Merck
All Photos(1)

Key Documents

8.52097

Sigma-Aldrich

Fmoc-Lys(biotin)-OH

≥95% (TLC), for peptide synthesis, Novabiochem®

Synonym(s):

Fmoc-Lys(biotin)-OH, N-α-Fmoc-N-ε-biotinyl-L-lysine

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€309.00
1 G
€595.00
8520970005
€2,430.00
8520970025
€2,670.00

€309.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€309.00
1 G
€595.00
8520970005
€2,430.00
8520970025
€2,670.00

About This Item

Empirical Formula (Hill Notation):
C31H38N4O6S
CAS Number:
Molecular Weight:
594.72
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

€309.00


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

Fmoc-Lys(biotin)-OH, Novabiochem®

Quality Level

product line

Novabiochem®

Assay

≥85.0% (HPLC)
≥95% (TLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

biotin

storage temp.

2-30°C

SMILES string

S1[C@H]([C@H]5NC(=O)N[C@H]5C1)CCCCC(=O)NCCCC[C@H](NC(=O)OCC2c3c(cccc3)c4c2cccc4)C(=O)O

InChI

1S/C31H38N4O6S/c36-27(15-6-5-14-26-28-25(18-42-26)33-30(39)35-28)32-16-8-7-13-24(29(37)38)34-31(40)41-17-23-21-11-3-1-9-19(21)20-10-2-4-12-22(20)23/h1-4,9-12,23-26,28H,5-8,13-18H2,(H,32,36)(H,34,40)(H,37,38)(H2,33,35,39)/t24-,25-,26-,28-/m0/s1

InChI key

OFIBQNGDYNGUEZ-OBXRUURASA-N

General description

A modified lysine derivative for the preparation of biotin-labeled peptides by Fmoc SPPS.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Biotinylation Reagents for Peptide Synthesis

Application

  • Supplemental Information for: Describes the reaction of amino functionalized resin with HCTU-activated Fmoc-Lys(biotin)-OH in DMF to install a biocytin molecule. Fmoc was deprotected by 20% (K Sapkota, JK Lucas, JW Faulkner, MF Lichte, YL Guo, tandf.figshare.com).

Linkage

Replaces: 04-12-1237

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 99.5 % (a/a)
Purity (TLC(CMA2)): ≥ 95 %
Assay (HPLC, area%): ≥ 85.0 % (a/a)
Solubility (0,1 mmole in 1 ml NMP): clearly soluble
Water (K. F.): ≤ 3.0 %
This product can contain up to 10% DMF in order to assure an ideal solubility.

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Biotin-labelled peptides find applications in immunology and histochemistry for affinity purification and receptor localization.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service