Skip to Content
Merck
All Photos(1)

Key Documents

8.52052

Sigma-Aldrich

Fmoc-His(Boc)-OH-CHA

Novabiochem®

Synonym(s):

Fmoc-His(Boc)-OH-CHA, N-α-Fmoc-N-im-t.-Boc-L-histidine cyclohexylammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H27N3O6 · xC6H13N
CAS Number:
Molecular Weight:
477.51
UNSPSC Code:
12352209

product line

Novabiochem®

Assay

≥97% (TLC)
≥97.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Boc
Fmoc

storage temp.

−20°C (−15°C to −25°C)

InChI

1S/C26H27N3O6.C6H13N/c1-26(2,3)35-25(33)29-13-16(27-15-29)12-22(23(30)31)28-24(32)34-14-21-19-10-6-4-8-17(19)18-9-5-7-11-20(18)21;7-6-4-2-1-3-5-6/h4-11,13,15,21-22H,12,14H2,1-3H3,(H,28,32)(H,30,31);6H,1-5,7H2/t22-;/m0./s1

InChI key

UDEXYAHZNOKVIG-FTBISJDPSA-N

General description

The N-im-Boc group is unstable to prolonged treatment with piperidine [1]; the utility of this derivative is therefore limited to the preparation of short sequences by Fmoc SPPS. This derivative is supplied as a CHA-salt.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Literature references

[1] P. Sieber, et al. (1987) Tetrahedron Lett., 28, 6031.

Linkage

Replaces: 04-12-1110

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 99.5 % (a/a)
Purity (TLC(011A)): ≥ 97 %
Purity (TLC(157B)): ≥ 97 %
Assay (HPLC, area%): ≥ 97.0 % (a/a)
Water (K. F.): ≤ 1.00 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service