Skip to Content
Merck
All Photos(1)

Key Documents

8.51093

Sigma-Aldrich

TDBA

4-(3-Trifluoromethyl)-3H-diazirin-3-yl)benzoic acid Novabiochem®

Synonym(s):

TDBA, 4-(3-(Trifluoromethyl)-3H-diazirin-3-yl)benzoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€315.00

€315.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€315.00

About This Item

Empirical Formula (Hill Notation):
C9H5F3N2O2
CAS Number:
Molecular Weight:
230.14
MDL number:
UNSPSC Code:
12352005
NACRES:
NA.22

€315.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

product line

Novabiochem®

form

powder

reaction suitability

reaction type: Photoclick reactions

manufacturer/tradename

Novabiochem®

mp

110-112 °C

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

FC(F)(F)[C@]2([N+H]=N2)c1ccc(cc1)C(=O)[O-]

InChI

1S/C9H5F3N2O2/c10-9(11,12)8(13-14-8)6-3-1-5(2-4-6)7(15)16/h1-4H,(H,15,16)

InChI key

CZPAJVBVULSLGG-UHFFFAOYSA-N

General description

A highly sensitive photocross-linker.activated at 300 nm for the preparation of photoactivatable biological probes [1].

Associated Protocols and Technical Articles
Guide to Selection of Coupling Reagents

Literature references

[1] M.Nassal, et al. ( 1983) Liebigs Ann. Chem.,, 1510.

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Assay (NMR-spectrum): ≥ 98.0 %

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service