Skip to Content
Merck
All Photos(1)

Key Documents

8.51016

Sigma-Aldrich

SAMA-OPfp

Novabiochem®

Synonym(s):

SAMA-OPfp, S-Acetylthioglycolic acid pentafluorophenyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H5F5O3S
CAS Number:
Molecular Weight:
300.20
MDL number:
UNSPSC Code:
12352108

product line

Novabiochem®

Assay

≥98% (TLC)

form

powder

reaction suitability

reagent type: ligand

manufacturer/tradename

Novabiochem®

mp

45-49 °C

application(s)

peptide synthesis

storage temp.

15-25°C

InChI

1S/C10H5F5O3S/c1-3(16)19-2-4(17)18-10-8(14)6(12)5(11)7(13)9(10)15/h2H2,1H3

InChI key

VAVNEUKAWUEHAG-UHFFFAOYSA-N

General description

This reagent [1] provides an effective means of linking synthetic peptide antigens to MAP core peptides [2] or carrier proteins for the purpose of raising antibodies [3]. Using this technique many of the problems associated with the analysis and purification of MAPs are avoided, since the linear peptide antigen can be fully characterized before conjugation to the preformed lysine tree.

The product number for this product was previously 01-63-0041.

To obtain a certificate of analysis (CoA) of a lot that begins with the letter “A”, please select the option in the right hand menu “Request a COA for Lot#s starting with A”.

Linkage

Replaces: 01-63-0041

Analysis Note

Colour (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Purity (TLC (0003)): ≥ 98 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service