Skip to Content
Merck
All Photos(1)

Documents

8.45001

Sigma-Aldrich

Acetaldehyde

Msynthplus

Synonym(s):

Acetaldehyde, Ethanal, Acetic Aldehyde, Ethyl Aldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H4O
CAS Number:
Molecular Weight:
44.05
MDL number:
UNSPSC Code:
12352114
EC Index Number:
200-836-8

vapor pressure

1006 hPa ( 20 °C)

Quality Level

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

140 °C

expl. lim.

4-57 % (v/v)

pH

5 (20 °C, 10 g/L in H2O)

bp

20.4 °C/1013 hPa

mp

-123 °C

transition temp

flash point -20 °C

density

0.78 g/cm3 at 20 °C

storage temp.

2-8°C

InChI

1S/C2H4O/c1-2-3/h2H,1H3

InChI key

IKHGUXGNUITLKF-UHFFFAOYSA-N

Analysis Note

Color (visual): colourless
Appearance of substance (visual): clear
Assay (GC, area%): ≥ 99.5 % (a/a)
Density (d 5°C/ 4°C): 0.799 - 0.801
Crotonaldehyde (GC, area%): ≤ 0.10 % (a/a)
Praldehyde (GC, area%): ≤ 0.10 % (a/a)
Acetic acid (GC, area%): ≤ 0.25 % (a/a)
Aldol (GC, area%): ≤ 0.10 % (a/a)
The analysis of residual solvents is not possible due to the physicochemical properties of the material.

Legal Information

MSYNTH is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 1 - Muta. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(C)

-38.89 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service