Skip to Content
Merck
All Photos(1)

Key Documents

8.43950

Sigma-Aldrich

Tetrahydropyran

for synthesis

Synonym(s):

Tetrahydropyran

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€78.90
1 L
€238.00

€78.90


Estimated to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
250 ML
€78.90
1 L
€238.00

About This Item

Empirical Formula (Hill Notation):
C5H10O
CAS Number:
Molecular Weight:
86.13
MDL number:
UNSPSC Code:
12352005
EC Index Number:
205-552-8
NACRES:
NA.22

€78.90


Estimated to ship on31 March 2025Details


Request a Bulk Order

Quality Level

Assay

≥99.0% (GC)

form

liquid

bp

88 °C/1013 hPa

mp

-49.2 °C

transition temp

flash point -16 °C

density

0.88 g/cm3 at 25 °C

storage temp.

2-30°C

SMILES string

O1CCCCC1

InChI

1S/C5H10O/c1-2-4-6-5-3-1/h1-5H2

InChI key

DHXVGJBLRPWPCS-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.882 - 0.885
Water (K. F.): ≤ 0.50 %
Identity (IR): passes test

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

3.2 °F - closed cup

Flash Point(C)

-16 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service