Skip to Content
Merck
All Photos(1)

Key Documents

8.43944

Sigma-Aldrich

Propanephosphonic acid anhydride

(50% solution in Dimethyl formamide) for synthesis T3P

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€69.10
100 ML
€185.00

€69.10


Estimated to ship on14 February 2025Details


Request a Bulk Order

Select a Size

Change View
25 ML
€69.10
100 ML
€185.00

About This Item

UNSPSC Code:
12352108
NACRES:
NA.22

€69.10


Estimated to ship on14 February 2025Details


Request a Bulk Order

Quality Level

form

liquid

bp

153 °C/1013 hPa (Dimethylformamide)

transition temp

flash point 58 °C (Dimethylformamide)

density

1.094 g/cm3

storage temp.

2-8°C

Analysis Note

Assay (T3P) (NMR): ≥ 40.0 %
Propanephosphonic acid (NMR): ≤ 0.50 %
Dimethylformamide (calculated): ≤ 50.0 %

Signal Word

Danger

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Repr. 1B - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

136.4 °F - (Dimethylformamide)

Flash Point(C)

58 °C - (Dimethylformamide)


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service