Skip to Content
Merck
All Photos(1)

Key Documents

8.43899

Sigma-Aldrich

Sodium molybdate dihydrate

for synthesis

Synonym(s):

Sodium molybdate dihydrate

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€38.10
250 G
€83.10

€38.10


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 G
€38.10
250 G
€83.10

About This Item

Empirical Formula (Hill Notation):
MoNa2O4 · 2H2O
CAS Number:
Molecular Weight:
241.95
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-551-7
NACRES:
NA.22

€38.10


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

crystals

potency

4233 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

pH

9-10 (20 °C, 840 g/L in H2O)

mp

687 °C (Elimination of water of crystallisation)

solubility

840 g/L

density

2.71 g/cm3 at 22 °C (for the dihydrate)

bulk density

1000‑1400 kg/m3

storage temp.

2-30°C

SMILES string

[Mo](=O)(=O)([O-])[O-].[Na+].[Na+]

InChI

1S/Mo.2Na.4O/q;2*+1;;;2*-1

InChI key

TVXXNOYZHKPKGW-UHFFFAOYSA-N

Related Categories

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service