Skip to Content
Merck
All Photos(1)

Key Documents

8.43487

Sigma-Aldrich

Manganese(II)acetate tetrahydrate

for synthesis

Synonym(s):

Manganese(II)acetate tetrahydrate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€41.90
250 G
€148.00

€41.90


Estimated to ship on03 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
€41.90
250 G
€148.00

About This Item

Linear Formula:
(CH3COO)2Mn · 4H2O
CAS Number:
Molecular Weight:
245.09
MDL number:
UNSPSC Code:
12352103
EC Index Number:
211-334-3
NACRES:
NA.22
grade:
for synthesis
synthesis grade
form:
solid
solubility:
233 g/L

€41.90


Estimated to ship on03 April 2025Details


Request a Bulk Order

grade

for synthesis
synthesis grade

Quality Level

vapor pressure

<1 hPa ( 25 °C)

form

solid

potency

3730 mg/kg LD50, oral (Rat)

pH

7.0 (20 °C, 50 g/L in H2O)

mp

80 °C

solubility

233 g/L

density

1.58 g/cm3 at 25 °C

bulk density

700 kg/m3

storage temp.

2-30°C

SMILES string

[Mn](OC(=O)C)OC(=O)C.O.O.O.O

InChI

1S/2C2H4O2.Mn.4H2O/c2*1-2(3)4;;;;;/h2*1H3,(H,3,4);;4*1H2/q;;+2;;;;/p-2

InChI key

CESXSDZNZGSWSP-UHFFFAOYSA-L

Analysis Note

Assay (complexometric): ≥ 99.0 %
Identity (IR): passes test

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3 - STOT RE 2 Inhalation

Target Organs

Brain

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service