Skip to Content
Merck
All Photos(1)

Key Documents

8.42128

Sigma-Aldrich

1,4-Diaminoanthraquinone

for synthesis

Synonym(s):

1,4-Diaminoanthraquinone

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€54.90
100 G
€136.00

€54.90


Estimated to ship on28 March 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
€54.90
100 G
€136.00

About This Item

Empirical Formula (Hill Notation):
C14H10N2O2
CAS Number:
Molecular Weight:
238.24
MDL number:
UNSPSC Code:
12352116
EC Index Number:
204-922-6
NACRES:
NA.22

€54.90


Estimated to ship on28 March 2025Details


Request a Bulk Order

vapor pressure

<1 hPa ( 25 °C)

Quality Level

Assay

≥88.0% (HPLC)

form

powder

autoignition temp.

>400 °C

potency

5790 mg/kg LD50, oral (Rat)

pH

7 (20 °C in H2O)

mp

268 °C

transition temp

flash point 340 °C

density

1.44 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

Nc1c2c(c(cc1)N)C(=O)c3c(cccc3)C2=O

InChI

1S/C14H10N2O2/c15-9-5-6-10(16)12-11(9)13(17)7-3-1-2-4-8(7)14(12)18/h1-6H,15-16H2

InChI key

FBMQNRKSAWNXBT-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 88.0 % (a/a)
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

644.0 °F - closed cup

Flash Point(C)

340 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service