Skip to Content
Merck
All Photos(1)

Key Documents

8.41790

Sigma-Aldrich

3,5-Dichloroaniline

for synthesis

Synonym(s):

3,5-Dichloroaniline

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€120.00

€120.00


Estimated to ship on24 March 2025Details



Select a Size

Change View
100 G
€120.00

About This Item

Empirical Formula (Hill Notation):
C6H5Cl2N
CAS Number:
Molecular Weight:
162.02
MDL number:
UNSPSC Code:
12352116
EC Index Number:
210-948-9
NACRES:
NA.22

€120.00


Estimated to ship on24 March 2025Details


vapor pressure

40 hPa ( 150 °C)

Quality Level

Assay

≥97.0% (GC)

autoignition temp.

620 °C

bp

259-260 °C/1013 hPa

mp

46-53 °C

transition temp

flash point 133 °C

density

1.58 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

Clc1cc(cc(c1)N)Cl

InChI

1S/C6H5Cl2N/c7-4-1-5(8)3-6(9)2-4/h1-3H,9H2

InChI key

UQRLKWGPEVNVHT-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Melting range (lower value): ≥ 46 °C
Melting range (upper value): ≤ 53 °C
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Target Organs

Kidney

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

271.4 °F - closed cup

Flash Point(C)

133 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service