Skip to Content
Merck
All Photos(1)

Key Documents

8.41603

Sigma-Aldrich

Dipropyl sulfide

for synthesis

Synonym(s):

Dipropyl sulfide

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€108.00

€108.00


Estimated to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
25 ML
€108.00

About This Item

Empirical Formula (Hill Notation):
C6H14S
CAS Number:
Molecular Weight:
118.24
MDL number:
UNSPSC Code:
12352113
EC Index Number:
203-873-8
NACRES:
NA.22

€108.00


Estimated to ship on31 March 2025Details


Request a Bulk Order

vapor pressure

8.6 hPa

Quality Level

form

liquid

expl. lim.

1-51 % (v/v)

bp

141-142 °C/1013 hPa

mp

-102 °C

transition temp

flash point 28 °C

density

0.838 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

S(CCC)CCC

InChI

1S/C6H14S/c1-3-5-7-6-4-2/h3-6H2,1-2H3

InChI key

ZERULLAPCVRMCO-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service