Skip to Content
Merck
All Photos(1)

Documents

8.41260

Sigma-Aldrich

1,1-Cyclobutanedicarboxylic acid

≥98%

Synonym(s):

1,1-Cyclobutanedicarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O4
CAS Number:
Molecular Weight:
144.13
MDL number:
UNSPSC Code:
12352106
EC Index Number:
226-651-2
NACRES:
NA.22

Quality Level

Assay

≥98%

form

powder

storage temp.

2-30°C

InChI

1S/C6H8O4/c7-4(8)6(5(9)10)2-1-3-6/h1-3H2,(H,7,8)(H,9,10)

InChI key

CCQPAEQGAVNNIA-UHFFFAOYSA-N

Related Categories

General description

for synthesis

Analysis Note

Assay (acidimetric): ≥ 98.0 %
Identity (IR): passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service