Skip to Content
Merck
All Photos(1)

Key Documents

8.41248

Sigma-Aldrich

3-Cyanopropyl-trichlorosilane

for synthesis

Synonym(s):

3-Cyanopropyl-trichlorosilane, 4-(Trichlorosilyl)-butyronitrile

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€137.00

€137.00


Estimated to ship on30 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 ML
€137.00

About This Item

Empirical Formula (Hill Notation):
C4H6Cl3NSi
CAS Number:
Molecular Weight:
202.54
MDL number:
UNSPSC Code:
12352103
EC Index Number:
213-990-6
NACRES:
NA.22

€137.00


Estimated to ship on30 April 2025Details


Request a Bulk Order

Quality Level

Assay

≥98.0% (GC)

form

liquid

potency

2830 mg/kg LD50, oral (Rat)

bp

237-238 °C/1013 hPa

transition temp

flash point 92 °C

density

1.28 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Si](Cl)(Cl)(Cl)CCCC#N

InChI

1S/C4H6Cl3NSi/c5-9(6,7)4-2-1-3-8/h1-2,4H2

InChI key

HMFFOEBLYHLRQN-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.301 - 1.305
Identity (IR): passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service