Skip to Content
Merck
All Photos(2)

Key Documents

8.40094

Sigma-Aldrich

(1R)-(-)-Camphor-10-sulfonic acid

for synthesis

Synonym(s):

(1R)-(-)-Camphor-10-sulfonic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€166.00

€166.00


Estimated to ship on02 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 G
€166.00

About This Item

Empirical Formula (Hill Notation):
C10H16O4S
CAS Number:
Molecular Weight:
232.30
MDL number:
UNSPSC Code:
12352106
EC Index Number:
252-817-9
NACRES:
NA.22

€166.00


Estimated to ship on02 April 2025Details


Request a Bulk Order

Quality Level

Assay

≥97.0% (acidimetric)

form

powder

pH

1.2-1.4 (20 g/L in H2O)

mp

193-196 °C (decomposes)

storage temp.

2-30°C

SMILES string

[S](=O)(=O)(O)C[C@@]21C(C(CC2)CC1=O)(C)C

InChI

1S/C10H16O4S/c1-9(2)7-3-4-10(9,8(11)5-7)6-15(12,13)14/h7H,3-6H2,1-2H3,(H,12,13,14)/t7?,10-/m0/s1

InChI key

MIOPJNTWMNEORI-MHPPCMCBSA-N

Analysis Note

Assay (acidimetric): ≥ 97.0 %
Melting range (lower value): ≥ 193 °C
Melting range (upper value): ≤ 196 °C
Spec. rotation [α²0/D (c=20 in water): -23.0 - -21.0 °
Identity (IR): passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service