Skip to Content
Merck
All Photos(1)

Key Documents

8.40056

Sigma-Aldrich

4,4′-Difluorodiphenylmethane

for synthesis

Synonym(s):

4,4′-Difluorodiphenylmethane, Bis(4-fluorophenyl)methane

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€41.50

€41.50


Estimated to ship on24 March 2025Details


Request a Bulk Order

Select a Size

Change View
10 G
€41.50

About This Item

CAS Number:
Molecular Weight:
204.22
MDL number:
UNSPSC Code:
12352101
EC Index Number:
207-274-2
NACRES:
NA.22

€41.50


Estimated to ship on24 March 2025Details


Request a Bulk Order

Quality Level

form

liquid

bp

265 °C

density

1.16 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

Fc1ccc(cc1)Cc2ccc(cc2)F

InChI

1S/C13H10F2/c14-12-5-1-10(2-6-12)9-11-3-7-13(15)8-4-11/h1-8H,9H2

InChI key

DXQVFHQUHOFROC-UHFFFAOYSA-N

General description

for synthesis

Application

4,4′-Difluorodiphenylmethane for synthesis. CAS 457-68-1, molar mass 204.22 g/mol.

Analysis Note

Assay (GC, area%)≥ 98.0 % (a/a)Density (d 20 °C/ 4 °C)1.157 - 1.161Identity (IR)passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service