Skip to Content
Merck
All Photos(2)

Key Documents

8.22324

Supelco

1,2-Propanediol

EMPLURA®

Synonym(s):

1,2-Propanediol, 1,2-Propylene glycol

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€60.10
5 L
€223.00

€60.10


Available to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
1 L
€60.10
5 L
€223.00

About This Item

Linear Formula:
CH3CH(OH)CH2OH
CAS Number:
Molecular Weight:
76.09
MDL number:
UNSPSC Code:
12352104
EC Index Number:
200-338-0
NACRES:
NB.21
bp:
188 °C/1013 hPa
vapor pressure:
0.11 hPa ( 20 °C)

€60.10


Available to ship on31 March 2025Details


Request a Bulk Order

vapor pressure

0.11 hPa ( 20 °C)

Quality Level

product line

EMPLURA®

Assay

≥99% (GC)

form

liquid

autoignition temp.

371 °C

potency

19400-36000 mg/kg LD50, oral (Rat)
20800 mg/kg LD50, skin (Rabbit)

expl. lim.

2.4-17.4 % (v/v)

dilution

(low-cost alternative, for basic applications)

impurities

≤0.2% Water

refractive index

n20/D 1.43

pH

6-8 (20 °C, 100 g/L in H2O)

bp

188 °C/1013 hPa

mp

-59 °C

transition temp

flash point 99 °C

density

1.04 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OC(CO)C

InChI

1S/C3H8O2/c1-3(5)2-4/h3-5H,2H2,1H3

InChI key

DNIAPMSPPWPWGF-UHFFFAOYSA-N

Application


  • (1)H-NMR metabolomics investigation of CSF from children with HIV reveals altered neuroenergetics due to persistent immune activation.: This research uses 1,2-Propanediol in metabolic profiling to study CSF in children with HIV, identifying neuroenergetic alterations due to persistent immune activation (Thirion et al., 2024).

  • Insight into the influence of natural deep eutectic solvents on the extraction of phenolic compounds from poplar type propolis: Composition and in vitro biological activity.: The study explores using 1,2-Propanediol-based deep eutectic solvents for extracting phenolic compounds from propolis, assessing their composition and biological activity (Trusheva et al., 2024).

  • In Silico-Assisted Isolation of trans-Resveratrol and trans-e-Viniferin from Grapevine Canes and Their Sustainable Extraction Using Natural Deep Eutectic Solvents (NADES).: This publication discusses the application of 1,2-Propanediol in the green chemistry extraction of valuable compounds from grapevines using computational and experimental methods (Kiene et al., 2023).

  • Untargeted metabolomics provide new insights into the implication of Lactobacillus helveticus strains isolated from natural whey starter in methylglyoxal-mediated browning.: Research utilizing 1,2-Propanediol in metabolomic studies to understand the metabolic pathways influenced by Lactobacillus strains in dairy products (Galimberti et al., 2023).

  • Milk oligosaccharide-driven persistence of Bifidobacterium pseudocatenulatum modulates local and systemic microbial metabolites upon synbiotic treatment in conventionally colonized mice.: A study focusing on the role of 1,2-Propanediol in modulating microbial metabolites through synbiotic treatments, impacting gut microbiota and systemic health (Larke et al., 2023).


Analysis Note

Purity (GC): ≥ 99.0 %
Identity (IR): conforms
Density (d 20 °C/ 4 °C): 1.036 - 1.037
Water: ≤ 0.20 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service