Skip to Content
Merck
All Photos(1)

Key Documents

8.22315

Sigma-Aldrich

1H-Benzotriazole

for synthesis

Synonym(s):

1H-Benzotriazole

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€47.30
500 G
€140.00
1 KG
€213.00
25 KG
€1,800.00

€47.30


Estimated to ship on05 May 2025Details


Request a Bulk Order

Select a Size

Change View
100 G
€47.30
500 G
€140.00
1 KG
€213.00
25 KG
€1,800.00

About This Item

Empirical Formula (Hill Notation):
C6H5N3
CAS Number:
Molecular Weight:
119.12
MDL number:
UNSPSC Code:
12352005
EC Index Number:
202-394-1
NACRES:
NA.22

€47.30


Estimated to ship on05 May 2025Details


Request a Bulk Order

vapor pressure

0.053 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (HPLC)

form

solid

autoignition temp.

400 °C

potency

560 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

pH

6.0-7.0 (20 °C, 100 g/L in H2O, suspension)

bp

350 °C/1013 hPa

mp

99 °C

transition temp

flash point 212 °C

density

1.36 g/cm3 at 20 °C

bulk density

500 kg/m3

storage temp.

2-30°C

SMILES string

[nH]1nc2c(n1)cccc2

InChI

1S/C6H5N3/c1-2-4-6-5(3-1)7-9-8-6/h1-4H,(H,7,8,9)

InChI key

QRUDEWIWKLJBPS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • A copper (I) coordination polymer incorporating the corrosion inhibitor 1H-benzotriazole: This study presents a novel copper (I) coordination polymer synthesized using 1H-benzotriazole, offering potential applications in corrosion inhibition (Liu et al., 2024).
  • Corrosion Behavior of Aluminum-Based Materials in Polymer Electrolyte Membrane Fuel Cell Environment: This paper investigates the effect of methyl-1H-benzotriazole on the corrosion behavior of aluminum, shedding light on its potential in enhancing the durability of fuel cell components (Jahan, 2024).
  • Electrocatalytic detection of Hg (II) using a platinum electrode modified with composite film: The incorporation of 1H-benzotriazole in a novel metallocyclic complex enhances its utility in the electrocatalytic detection of mercury, suggesting applications in environmental monitoring (Shoba et al., 2024).

Analysis Note

Assay (HPLC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 95 °C
Melting range (upper value): ≤ 99 °C
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

338.0 °F - closed cup

Flash Point(C)

170 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service