Skip to Content
Merck
All Photos(1)

Key Documents

8.22312

Sigma-Aldrich

4-Aminobenzoic acid

≥99.0% (acidimetric)

Synonym(s):

4-Aminobenzoic acid, PABA

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€57.20
1 KG
€212.00
25 KG
€1,310.00

€57.20


Estimated to ship on05 May 2025Details


Request a Bulk Order

Select a Size

Change View
250 G
€57.20
1 KG
€212.00
25 KG
€1,310.00

About This Item

Empirical Formula (Hill Notation):
C7H7NO2
CAS Number:
Molecular Weight:
137.14
MDL number:
UNSPSC Code:
12352106
EC Index Number:
205-753-0
NACRES:
NA.22

€57.20


Estimated to ship on05 May 2025Details


Request a Bulk Order

Product Name

4-Aminobenzoic acid, for synthesis

Quality Level

Assay

≥99.0% (acidimetric)

form

solid

potency

6000 mg/kg LD50, oral (Rat)

pH

3.5 (20 °C, 5 g/L in H2O)

mp

186-189 °C

density

1.38 g/cm3 at 20 °C

bulk density

200 kg/m3

storage temp.

2-30°C

SMILES string

[N+H3]c1ccc(cc1)C(=O)[O-]

InChI

1S/C7H7NO2/c8-6-3-1-5(2-4-6)7(9)10/h1-4H,8H2,(H,9,10)

InChI key

ALYNCZNDIQEVRV-UHFFFAOYSA-N

Application

  • Synthesis of Yellow Fluorescence Carbon Dots: 4-Aminobenzoic acid is used to produce yellow fluorescence carbon dots, highlighting its application in vitamin B(12) detection and cell imaging (Zhao et al., 2023).
  • Enhanced Production of 4-Amino-3-Hydroxybenzoic Acid: 4-hydroxybenzoate 3-hydroxylase was engineered to convert glucose into 4-amino-3-hydroxybenzoic acid in Corynebacterium glutamicum, illustrating a biotechnological application of 4-Aminobenzoic acid (Nonaka et al., 2023).
  • Electrochemical Sensor Development: The compound was used to functionalize diazonium salt modified screen-printed graphene electrodes, creating a rapid diagnostic tool for detecting interleukin-6 (IL-6), relevant in sepsis diagnosis (Jesadabundit et al., 2023).

Analysis Note

Assay (acidimetric): ≥ 99.0 %
Melting range (lower value): ≥ 185 °C
Melting range (upper value): ≤ 188 °C
Identity (IR): passes test

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

339.8 °F

Flash Point(C)

171 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service