Skip to Content
Merck
All Photos(1)

Key Documents

8.22292

Sigma-Aldrich

4-Nitroaniline

for synthesis

Synonym(s):

4-Nitroaniline, p-Nitraniline

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€42.80
1 KG
€119.00

€42.80


Estimated to ship on07 May 2025Details


Request a Bulk Order

Select a Size

Change View
250 G
€42.80
1 KG
€119.00

About This Item

Empirical Formula (Hill Notation):
C6H6N2O2
CAS Number:
Molecular Weight:
138.12
MDL number:
UNSPSC Code:
12352116
EC Index Number:
202-810-1
NACRES:
NA.22

€42.80


Estimated to ship on07 May 2025Details


Request a Bulk Order

vapor pressure

1.33 hPa ( 142 °C)

Quality Level

Assay

≥98.0% (GC)

form

solid

autoignition temp.

500 °C

bp

>331 °C/1013 hPa (decomposition)

mp

148 °C

transition temp

flash point 199 °C

solubility

0.5 g/L

density

1.44 g/cm3 at 20 °C

bulk density

630 kg/m3

storage temp.

2-30°C

SMILES string

[N+](=O)([O-])c1ccc(cc1)N

InChI

1S/C6H6N2O2/c7-5-1-3-6(4-2-5)8(9)10/h1-4H,7H2

InChI key

TYMLOMAKGOJONV-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 146 °C
Melting range (upper value): ≤ 150 °C
Identity (IR): passes test

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - STOT RE 2

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

415.4 °F - closed cup

Flash Point(C)

213.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service